COM456878716 Combi-Blocks, Inc.

Ethyl 1-cyclopentyl-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD28962784
  • Cas Number : 1845690-60-9
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9335

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9335
Availability details for
Quantity
  • Availability
  • Add to Cart