COM456878717 Combi-Blocks, Inc.

Ethyl 1-cyclopentyl-1,2,3-benzotriazole-5-carboxylate

  • MDL number MFCD28962785
  • Cas Number : 1845706-36-6
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9337
Availability details for
Quantity
  • Availability
  • Add to Cart