COM456878722 Combi-Blocks, Inc.

4-Chloro-6-(trifluoromethyl)-1,2,3-benzotriazole

  • MDL number MFCD28962790
  • Cas Number : 1086836-70-5
  • Molecular Weight : 221.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9402

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9402
Availability details for
Quantity
  • Availability
  • Add to Cart