COM456878728 Combi-Blocks, Inc.

8-Bromo-6-fluoro-1-methyl-4H-quinoxaline-2,3-dione

  • MDL number MFCD28515416
  • Cas Number : 1845717-02-3
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9836

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9836
Availability details for
Quantity
  • Availability
  • Add to Cart