COM456878732 Combi-Blocks, Inc.

4-Bromo-2-methanesulfonyl-N-methylaniline

  • MDL number MFCD28962797
  • Cas Number : 1845690-63-2
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9912

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9912
Availability details for
Quantity
  • Availability
  • Add to Cart