COM456878736 Combi-Blocks, Inc.

2-Benzoylpyrrolidine hydrochloride

  • MDL number MFCD16295304
  • Cas Number : 138371-64-9
  • Molecular Weight : 211.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4232

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4232
Availability details for
Quantity
  • Availability
  • Add to Cart