COM456878745 Combi-Blocks, Inc.

2,6-Difluoro-N,N-dimethylbenzamide

  • MDL number MFCD20356572
  • Cas Number : 81652-57-5
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4283

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4283
Availability details for
Quantity
  • Availability
  • Add to Cart