COM456878753 Combi-Blocks, Inc.

4-Bromo-1-(4-chlorophenyl)pyrazole

  • MDL number MFCD17677464
  • Cas Number : 885017-82-3
  • Molecular Weight : 257.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2185

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2185
Availability details for
Quantity
  • Availability
  • Add to Cart