COM456878756 Combi-Blocks, Inc.

1-(Triisopropylsilyl)pyrrole-3-carbaldehyde

  • MDL number MFCD28515390
  • Cas Number : 90971-76-9
  • Molecular Weight : 251.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2222

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2222
Availability details for
Quantity
  • Availability
  • Add to Cart