COM456878757 Combi-Blocks, Inc.

5-Benzoyl-4H,6H,7H-[1,3]thiazolo[5,4-c]pyridin-2-amine

  • MDL number MFCD10010952
  • Cas Number : 17899-57-9
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2226

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2226
Availability details for
Quantity
  • Availability
  • Add to Cart