COM456878758 Combi-Blocks, Inc.

Methyl 4-(pyrrol-1-yl)benzoate

  • MDL number MFCD06010008
  • Cas Number : 23351-08-8
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-(1H-pyrrol-1-yl)benzoate
Inchi Code 1S/C12H11NO2/c1-15-12(14)10-4-6-11(7-5-10)13-8-2-3-9-13/h2-9H,1H3
Inchi Key XCBRROUJSCMUGF-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2227
Availability details for
Quantity
  • Availability
  • Add to Cart