COM456878768 Combi-Blocks, Inc.

2,3-Dioxo-1H-indole-5-sulfonyl chloride

  • MDL number MFCD11974220
  • Cas Number : 132898-96-5
  • Molecular Weight : 245.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0526

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0526
Availability details for
Quantity
  • Availability
  • Add to Cart