COM456878772 Combi-Blocks, Inc.

N-(2-(morpholinoethyl)-4-nitroaniline

  • MDL number MFCD11116216
  • Cas Number : 77755-81-8
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1206

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1206
Availability details for
Quantity
  • Availability
  • Add to Cart