COM456878774 Combi-Blocks, Inc.

tert-Butyl N-(5-methyl-1,3-thiazol-2-yl)carbamate

  • MDL number MFCD20039782
  • Cas Number : 1186298-92-9
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1227
Availability details for
Quantity
  • Availability
  • Add to Cart