COM456878793 Combi-Blocks, Inc.

5-Chloro-1,3-benzodioxole

  • MDL number MFCD00010842
  • Cas Number : 7228-38-8
  • Molecular Weight : 156.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-2087

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-2087
Availability details for
Quantity
  • Availability
  • Add to Cart