COM456878798 Combi-Blocks, Inc.

4-(N-Butoxy)benzenesulfonyl chloride

  • MDL number MFCD00052344
  • Cas Number : 1138-56-3
  • Molecular Weight : 248.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3200

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3200
Availability details for
Quantity
  • Availability
  • Add to Cart