COM456878831 Combi-Blocks, Inc.

(1-Benzothiophen-4-yl)boronic acid

  • MDL number MFCD22191951
  • Cas Number : 177735-30-7
  • Molecular Weight : 178.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1098

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1098
Availability details for
Quantity
  • Availability
  • Add to Cart