COM456878833 Combi-Blocks, Inc.

(2,3-Dichloro-4-formylphenyl)boronic acid

  • MDL number MFCD18968536
  • Cas Number : 352535-89-8
  • Molecular Weight : 218.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1101

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1101
Availability details for
Quantity
  • Availability
  • Add to Cart