COM456878843 Combi-Blocks, Inc.

2,5-Difluoro-4-nitroaniline

  • MDL number MFCD11519059
  • Cas Number : 1542-36-5
  • Molecular Weight : 174.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5-difluoro-4-nitroaniline
Inchi Code 1S/C6H4F2N2O2/c7-3-2-6(10(11)12)4(8)1-5(3)9/h1-2H,9H2
Inchi Key IBGGCVMFSJQOID-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3538

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3538
Availability details for
Quantity
  • Availability
  • Add to Cart