COM456878855 Combi-Blocks, Inc.

1-Cyclopentyl-4-(4-nitrophenyl)piperazine

  • MDL number MFCD10040128
  • Cas Number : 443915-54-6
  • Molecular Weight : 275.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3617

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3617
Availability details for
Quantity
  • Availability
  • Add to Cart