COM456878858 Combi-Blocks, Inc.

4-(2-Methylphenoxy)benzonitrile

  • MDL number MFCD02068124
  • Cas Number : 24789-50-2
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4064

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4064
Availability details for
Quantity
  • Availability
  • Add to Cart