COM456878860 Combi-Blocks, Inc.

4-Bromo-2-(4-chlorophenoxymethyl)-1-methoxybenzene

  • MDL number MFCD19444754
  • Cas Number : 1285022-19-6
  • Molecular Weight : 327.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4070
Availability details for
Quantity
  • Availability
  • Add to Cart