COM456878862 Combi-Blocks, Inc.

1-Chloro-2-(chloromethyl)-4-methylbenzene

  • MDL number MFCD12025216
  • Cas Number : 34896-68-9
  • Molecular Weight : 175.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 96

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4072
Availability details for
Quantity
  • Availability
  • Add to Cart