COM456878870 Combi-Blocks, Inc.

2-[(4-Bromo-2-nitrophenyl)(methyl)amino]ethanol

  • MDL number MFCD14586990
  • Cas Number : 1249593-20-1
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6038

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6038
Availability details for
Quantity
  • Availability
  • Add to Cart