COM456878871 Combi-Blocks, Inc.

2-Chloro-3-ethoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD22564309
  • Cas Number : 2223029-21-6
  • Molecular Weight : 283.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1112

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1112
Availability details for
Quantity
  • Availability
  • Add to Cart