COM456878874 Combi-Blocks, Inc.

2-Bromo-3-fluoro-6-nitrotoluene

  • MDL number MFCD23704111
  • Cas Number : 1427502-92-8
  • Molecular Weight : 234.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6052

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6052
Availability details for
Quantity
  • Availability
  • Add to Cart