COM456878876 Combi-Blocks, Inc.

2-Chloro-3-propoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD26404836
  • Molecular Weight : 297.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1117

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1117
Availability details for
Quantity
  • Availability
  • Add to Cart