COM456878878 Combi-Blocks, Inc.

[3-(aminomethyl)-5-fluorophenyl]boronic acid

  • MDL number MFCD22397988
  • Molecular Weight : 169.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1138

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1138
Availability details for
Quantity
  • Availability
  • Add to Cart