COM456878882 Combi-Blocks, Inc.

2-(3-Aminophenyl)acetamide hydrochloride

  • MDL number MFCD28962746
  • Cas Number : 1845693-84-6
  • Molecular Weight : 186.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6079

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6079
Availability details for
Quantity
  • Availability
  • Add to Cart