COM456878884 Combi-Blocks, Inc.

2-Chloro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-ol

  • MDL number MFCD18726696
  • Cas Number : 2000236-32-6
  • Molecular Weight : 255.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1130

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1130
Availability details for
Quantity
  • Availability
  • Add to Cart