COM456878889 Combi-Blocks, Inc.

4-[(tert-Butyldimethylsilyl)oxy]-2,6-dimethylphenylboronic acid pinacol ester

  • MDL number MFCD22493751
  • Cas Number : 955930-05-9
  • Molecular Weight : 362.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1159
Availability details for
Quantity
  • Availability
  • Add to Cart