COM456878890 Combi-Blocks, Inc.

4-Methanesulfonyl-2-methylphenylboronic acid pinacol ester

  • MDL number MFCD18732808
  • Cas Number : 1011459-26-9
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0480

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-0480
Availability details for
Quantity
  • Availability
  • Add to Cart