COM456878891 Combi-Blocks, Inc.

2-Carboxy-5-chlorophenylboronic acid pinacol ester

  • MDL number MFCD24039757
  • Cas Number : 2377606-42-1
  • Molecular Weight : 282.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2747

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2747
Availability details for
Quantity
  • Availability
  • Add to Cart