COM456878892 Combi-Blocks, Inc.

5-Carboxy-2-fluorophenylboronic acid pinacol ester

  • MDL number MFCD23098802
  • Cas Number : 2235384-35-5
  • Molecular Weight : 266.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2883

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2883
Availability details for
Quantity
  • Availability
  • Add to Cart