COM456878895 Combi-Blocks, Inc.

4-[(tert-Butyldimethylsilyl)oxy]-2,6-dimethylphenylboronic acid

  • MDL number MFCD28009320
  • Cas Number : 858096-68-1
  • Molecular Weight : 280.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1159
Availability details for
Quantity
  • Availability
  • Add to Cart