COM456878933 Combi-Blocks, Inc.

2-Bromo-4-fluorobenzenesulfonyl chloride

  • MDL number MFCD03094149
  • Cas Number : 351003-45-7
  • Molecular Weight : 273.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5181

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5181
Availability details for
Quantity
  • Availability
  • Add to Cart