COM456878983 Combi-Blocks, Inc.

4-Fluoro-2,3-dimethylbenzoic acid

  • MDL number MFCD23712108
  • Cas Number : 1104380-03-1
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-4984

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-4984
Availability details for
Quantity
  • Availability
  • Add to Cart