COM456878984 Combi-Blocks, Inc.

5-Bromo-4-methoxy-2-(trifluoromethyl)pyridine

  • MDL number MFCD18257659
  • Cas Number : 1256790-33-6
  • Molecular Weight : 256.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2053

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2053
Availability details for
Quantity
  • Availability
  • Add to Cart