COM456878987 Combi-Blocks, Inc.

5-Bromo-2-fluoro-3-methylbenzonitrile

  • MDL number MFCD25955533
  • Cas Number : 1110502-49-2
  • Molecular Weight : 214.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5782

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5782
Availability details for
Quantity
  • Availability
  • Add to Cart