COM456878996 Combi-Blocks, Inc.

Ethyl 5-(pyridin-3-yl)-1,2-oxazole-3-carboxylate

  • MDL number MFCD08701226
  • Cas Number : 215870-77-2
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2099

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2099
Availability details for
Quantity
  • Availability
  • Add to Cart