COM456878999 Combi-Blocks, Inc.

(2,6-Dichloro-5-fluoropyridin-3-yl)methanamine

  • MDL number MFCD06213012
  • Cas Number : 771581-97-6
  • Molecular Weight : 195.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2103

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2103
Availability details for
Quantity
  • Availability
  • Add to Cart