COM456879002 Combi-Blocks, Inc.

Methyl 5-(benzyloxy)-2-bromobenzoate

  • MDL number MFCD21603891
  • Cas Number : 431052-32-3
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5815

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5815
Availability details for
Quantity
  • Availability
  • Add to Cart