COM456879003 Combi-Blocks, Inc.

Ethyl (2e)-3-(2-bromopyridin-3-yl)acrylate

  • MDL number MFCD26517231
  • Cas Number : 1715957-78-0
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2107
Availability details for
Quantity
  • Availability
  • Add to Cart