COM456879007 Combi-Blocks, Inc.

methyl 2-(2-bromo-6-fluorophenyl)acetate

  • MDL number MFCD22988859
  • Cas Number : 1427397-15-6
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5819

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5819
Availability details for
Quantity
  • Availability
  • Add to Cart