COM456879014 Combi-Blocks, Inc.

Methyl 3-(3-fluorophenyl)propanoate

  • MDL number MFCD12031749
  • Cas Number : 425704-52-5
  • Molecular Weight : 182.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-(3-fluorophenyl)propanoate
Inchi Code 1S/C10H11FO2/c1-13-10(12)6-5-8-3-2-4-9(11)7-8/h2-4,7H,5-6H2,1H3
Inchi Key ZZUWMASGVAWMSC-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5822

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5822
Availability details for
Quantity
  • Availability
  • Add to Cart