COM456879036 Combi-Blocks, Inc.

4-Methyl-3-(methylsulfonyl)benzoic acid

  • MDL number MFCD10032483
  • Cas Number : 51522-22-6
  • Molecular Weight : 214.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5359

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5359
Availability details for
Quantity
  • Availability
  • Add to Cart