COM456879040 Combi-Blocks, Inc.

Potassium trifluoro(3-fluoro-4-methylphenyl)borate

  • MDL number MFCD09993124
  • Molecular Weight : 216.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?TB-2209

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?TB-2209
Availability details for
Quantity
  • Availability
  • Add to Cart