COM456879043 Combi-Blocks, Inc.

3-Bromo-4-methylbenzenesulfonyl chloride

  • MDL number MFCD12922946
  • Cas Number : 1029145-99-0
  • Molecular Weight : 269.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9240

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9240
Availability details for
Quantity
  • Availability
  • Add to Cart