COM456879044 Combi-Blocks, Inc.

(S,S)-3-Isopropyl-6-methylpiperazine-2,5-dione

  • MDL number MFCD24549359
  • Cas Number : 15136-26-2
  • Molecular Weight : 170.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2420

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2420
Availability details for
Quantity
  • Availability
  • Add to Cart