COM456879045 Combi-Blocks, Inc.

Ethyl (2Z)-2-hydroxy-4-oxo-4-(pyridin-3-yl)but-2-enoate

  • MDL number MFCD26684187
  • Cas Number : 778531-36-5
  • Molecular Weight : 221.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5826

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5826
Availability details for
Quantity
  • Availability
  • Add to Cart